The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284230.1 | NP_001271159.1 | 6 - 7 | 1567 | 71 |
NM_001284231.1 | NP_001271160.1 | 6 - 7 | 1023 | 71 | |
NM_001284232.1 | NP_001271161.1 | 5 - 6 | 977 | 71 | |
NM_033141.3 | NP_149132.2 | 6 - 7 | 1567 | 71 | |
XM_005267683.4 | XP_005267740.1 | 6 - 7 | 1662 | 71 | |
XM_011536788.2 | XP_011535090.1 | 6 - 7 | 1664 | 71 | |
XM_011536792.1 | XP_011535094.1 | 5 - 6 | 977 | 71 | |
XM_011536794.2 | XP_011535096.1 | 4 - 5 | 2324 | 71 | |
GenBank mRNA | AF251442.1 | - | 6 - 7 | 1412 | 71 |
AK294571.1 | - | 5 - 6 | 948 | 71 | |
AK302849.1 | - | 6 - 7 | 1016 | 71 | |
AY327900.1 | - | 6 - 7 | 1567 | 71 | |
BC111407.1 | - | 6 - 7 | 1566 | 71 | |
BC133706.1 | - | 6 - 7 | 1567 | 71 | |
BX648924.1 | - | 5 - 6 | 965 | 71 |