The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002917.1 | NP_002908.1 | 6 - 7 | 839 | 115 |
XM_011523587.2 | XP_011521889.1 | 5 - 6 | 533 | 115 | |
GenBank mRNA | AF038196.1 | - | 3 - 4 | 413 | 115 |
AK094016.1 | - | 3 - 4 | 1054 | 115 | |
AK293568.1 | - | 7 - 8 | 785 | 115 | |
AK302068.1 | - | 6 - 7 | 648 | 115 | |
BC014495.1 | - | 2 - 3 | 287 | 115 | |
BC069034.1 | - | 2 - 3 | 766 | 115 | |
BC146805.1 | - | 3 - 4 | 1895 | 115 | |
U94353.1 | - | 3 - 4 | 411 | 115 |