Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERMARD
Entrez Gene ID
55780
Gene Name
ER membrane associated RNA degradation
Gene Aliases
C6orf70, PVNH6, dJ266L20.3
Chromosome Location
Chr.6: 169751622 - 169781584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278531.1 NP_001265460.1 14 - 15 1426 69
NM_001278532.1 NP_001265461.1 13 - 14 1257 69
NM_001278533.1 NP_001265462.1 14 - 15 1426 69
NM_018341.2 NP_060811.1 14 - 15 1426 69
XM_011535938.2 XP_011534240.1 13 - 14 1396 69
XM_011535939.2 XP_011534241.1 14 - 15 2516 69
XM_011535940.2 XP_011534242.1 14 - 15 2325 69
XM_017011027.1 XP_016866516.1 14 - 15 2266 69
XM_017011028.1 XP_016866517.1 14 - 15 1591 69
XM_017011029.1 XP_016866518.1 14 - 15 1351 69
XM_017011030.1 XP_016866519.1 13 - 14 1175 69
XM_017011031.1 XP_016866520.1 13 - 14 1240 69
XM_017011032.1 XP_016866521.1 14 - 15 2511 69
XM_017011033.1 XP_016866522.1 14 - 15 2320 69
GenBank mRNA AK001369.1 - 13 - 14 1287 69
AK002014.1 - 14 - 15 1423 69
AK021520.1 - 4 - 5 1490 69
AK294090.1 - 13 - 14 1254 69
BC044812.1 - 14 - 15 1410 69

View Full Product Details