The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010852.3 | NP_001010852.2 | 3 - 4 | 1898 | 107 |
XM_017010266.1 | XP_016865755.1 | 3 - 4 | 3600 | 107 | |
GenBank mRNA | AK095527.1 | - | 3 - 4 | 1902 | 107 |
AK295158.1 | - | 3 - 4 | 360 | 107 | |
BC101376.1 | - | 3 - 4 | 1432 | 107 | |
BC101377.2 | - | 3 - 4 | 1434 | 107 |