The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020440.3 | NP_065173.2 | 7 - 8 | 2449 | 118 |
XM_017001874.1 | XP_016857363.1 | 7 - 8 | 2220 | 118 | |
GenBank mRNA | AB014734.1 | - | 1 - 2 | 89 | 118 |
AB037857.1 | - | 7 - 8 | 2305 | 118 | |
AK074637.1 | - | 7 - 8 | 2315 | 118 | |
BC098142.1 | - | 7 - 8 | 1895 | 118 | |
BC114520.1 | - | 6 - 7 | 1746 | 118 | |
BC139718.1 | - | 6 - 7 | 1492 | 118 | |
BC152454.1 | - | 7 - 8 | 2305 | 118 |