The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130864.1 | NP_001124336.1 | 1 - 2 | 638 | 140 |
NM_001267035.1 | NP_001253964.1 | 1 - 2 | 638 | 140 | |
NM_052927.2 | NP_443159.1 | 1 - 2 | 638 | 140 | |
XM_011534424.2 | XP_011532726.1 | 1 - 2 | 678 | 140 | |
XM_017009012.1 | XP_016864501.1 | 1 - 2 | 677 | 140 | |
GenBank mRNA | AB067522.1 | - | 1 - 2 | 599 | 140 |
BC035076.2 | - | 1 - 2 | 616 | 140 | |
BC105110.1 | - | 1 - 2 | 615 | 140 | |
BC143437.1 | - | 1 - 2 | 613 | 140 |