The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027688.2 | 1 - 2 | 125 | 62 | |
GenBank mRNA | AK055955.1 | - | 1 - 2 | 125 | 62 |