Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKR1C6P
Entrez Gene ID
389932
Gene Name
aldo-keto reductase family 1 member C6, pseudogene
Gene Aliases
TAKR
Chromosome Location
Chr.10: 4871667 - 4916273 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_026743.1 4 - 5 448 71
GenBank mRNA AB037902.1 - 2 - 3 251 71
CB111913.1 - 4 - 5 403 71
DA643129.1 - 3 - 4 381 71


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream aldo-keto reductase family 1 member E2 AKR1E2 AKR1CL2, AKRDC1, LoopADR, hTSP, htAKR -34.0kb Anti-sense Chr. - 4824554 - 4908421 on Build GRCh38
Downstream aldo-keto reductase family 1 member C1 AKR1C1 2-ALPHA-HSD, 20-ALPHA-HSD, C9, DD1, DD1/DD2, DDH, DDH1, H-37, HAKRC, HBAB, MBAB 73.3kb Anti-sense Chr. - 4963262 - 4979967 on Build GRCh38

View Full Product Details