Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTSL5
Entrez Gene ID
339366
Gene Name
ADAMTS like 5
Gene Aliases
THSD6
Chromosome Location
Chr.19: 1505018 - 1513189 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213604.2 NP_998769.2 4 - 5 695 127
XM_005259549.3 XP_005259606.1 5 - 6 404 127
XM_011527961.1 XP_011526263.1 5 - 6 404 127
XM_011527963.1 XP_011526265.1 5 - 6 403 127
XM_017026744.1 XP_016882233.1 4 - 5 526 127
XM_017026745.1 XP_016882234.1 4 - 5 558 127
GenBank mRNA AK131571.1 - 4 - 5 526 127
AK302020.1 - 5 - 6 430 127
BC040620.1 - 4 - 5 695 127
BC119796.1 - 4 - 5 329 127
BC139833.1 - 4 - 5 289 127

View Full Product Details