Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 14489111 - 14602996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080423.3 NP_001073892.3 1 - 2 77 87
XM_011534146.2 XP_011532448.1 1 - 2 152 87
GenBank mRNA AB051506.2 - 1 - 2 63 87

View Full Product Details