Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS1
Entrez Gene ID
26173
Gene Name
integrator complex subunit 1
Gene Aliases
INT1, NET28
Chromosome Location
Chr.7: 1470277 - 1504382 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080453.2 NP_001073922.2 12 - 13 1810 55
XM_011515260.1 XP_011513562.1 12 - 13 1810 55
XM_011515262.2 XP_011513564.1 12 - 13 1810 55
XM_017011959.1 XP_016867448.1 12 - 13 1810 55
XM_017011960.1 XP_016867449.1 12 - 13 1810 55
GenBank mRNA BF346658.1 - 3 - 4 345 55
BX324321.2 - 2 - 3 292 55

View Full Product Details