The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080453.2 | NP_001073922.2 | 3 - 4 | 448 | 73 |
XM_011515260.1 | XP_011513562.1 | 3 - 4 | 448 | 73 | |
XM_011515262.2 | XP_011513564.1 | 3 - 4 | 448 | 73 | |
XM_017011959.1 | XP_016867448.1 | 3 - 4 | 448 | 73 | |
XM_017011960.1 | XP_016867449.1 | 3 - 4 | 448 | 73 | |
GenBank mRNA | AI935093.1 | - | 1 - 2 | 169 | 73 |
DB113520.1 | - | 3 - 4 | 448 | 73 |