The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136537.1 | NP_001130009.1 | 7 - 8 | 1078 | 67 |
XM_017000449.1 | XP_016855938.1 | 7 - 8 | 2411 | 67 | |
GenBank mRNA | AK056150.1 | - | 3 - 4 | 709 | 67 |