Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAF4
Entrez Gene ID
57466
Gene Name
SR-related CTD associated factor 4
Gene Aliases
SFRS15, SRA4
Chromosome Location
Chr.21: 31671000 - 31732118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145444.1 NP_001138916.1 11 - 12 1901 89
NM_001145445.1 NP_001138917.1 12 - 13 1946 89
NM_020706.2 NP_065757.1 12 - 13 1946 89
XM_005261017.2 XP_005261074.1 11 - 12 1750 89
XM_006724035.2 XP_006724098.1 12 - 13 1915 89
XM_006724036.2 XP_006724099.1 12 - 13 1915 89
XM_017028415.1 XP_016883904.1 12 - 13 1904 89
XM_017028416.1 XP_016883905.1 12 - 13 1789 89
XM_017028417.1 XP_016883906.1 11 - 12 1750 89
GenBank mRNA AB032998.1 - 6 - 7 934 89
AF023142.1 - 12 - 13 1550 89
AK025748.1 - 7 - 8 965 89
AK308406.1 - 11 - 12 1752 89
AL117417.1 - 7 - 8 954 89
AL834304.1 - 12 - 13 1726 89
BC014921.1 - 12 - 13 1923 89
BC043353.1 - 12 - 13 1903 89
BC052286.1 - 12 - 13 1777 89
BC064990.1 - 12 - 13 1777 89

View Full Product Details