Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL22A1
Entrez Gene ID
169044
Gene Name
collagen type XXII alpha 1
Gene Aliases
-
Chromosome Location
Chr.8: 138588235 - 138914087 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152888.2 NP_690848.1 63 - 64 5080 59
XM_011516883.2 XP_011515185.1 62 - 63 5098 59
XM_011516884.2 XP_011515186.1 61 - 62 5059 59
XM_011516885.2 XP_011515187.1 61 - 62 5044 59
XM_011516886.2 XP_011515188.1 63 - 64 5010 59
XM_011516887.2 XP_011515189.1 59 - 60 3978 59
XM_011516889.2 XP_011515191.1 52 - 53 3219 59
XM_017013150.1 XP_016868639.1 63 - 64 4971 59
XM_017013151.1 XP_016868640.1 59 - 60 4936 59
XM_017013152.1 XP_016868641.1 58 - 59 3968 59
GenBank mRNA AF406780.1 - 63 - 64 5067 59
BC042075.1 - 37 - 38 2305 59
BC144535.1 - 51 - 52 3738 59

View Full Product Details