The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002917.1 | NP_002908.1 | 2 - 3 | 323 | 86 |
GenBank mRNA | AK293568.1 | - | 3 - 4 | 269 | 86 |
AK302068.1 | - | 2 - 3 | 132 | 86 | |
BF435018.1 | - | 2 - 3 | 323 | 86 |