Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1GALT1
Entrez Gene ID
56913
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1
Gene Aliases
C1GALT, T-synthase
Chromosome Location
Chr.7: 7182547 - 7248651 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020156.4 NP_064541.1 2 - 3 442 106
XM_005249812.1 XP_005249869.1 2 - 3 510 106
XM_011515453.2 XP_011513755.1 3 - 4 639 106
XM_011515455.2 XP_011513757.1 3 - 4 561 106
XM_011515456.2 XP_011513758.1 2 - 3 373 106
XM_017012442.1 XP_016867931.1 3 - 4 561 106
XM_017012443.1 XP_016867932.1 3 - 4 556 106
XM_017012444.1 XP_016867933.1 4 - 5 547 106
XM_017012445.1 XP_016867934.1 5 - 6 762 106
XM_017012446.1 XP_016867935.1 2 - 3 391 106
XM_017012447.1 XP_016867936.1 3 - 4 485 106
XM_017012448.1 XP_016867937.1 2 - 3 396 106
GenBank mRNA AF155582.1 - 1 - 2 281 106
AJ132443.1 - 1 - 2 219 106
AJ243256.1 - 1 - 2 219 106
AJ278960.1 - 1 - 2 219 106
DA977378.1 - 2 - 3 442 106

View Full Product Details