Assay Design

The assay probe spans an exon junction.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK095353.1 - 1 - 2 1099 Chr. X - 136840932 - 136847798 [-] on Build GRCh38 100


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream Rac/Cdc42 guanine nucleotide exchange factor 6 ARHGEF6 COOL2, Cool-2, MRX46, PIXA, alpha-PIX, alphaPIX -59.3kb Sense Chr. - 136665547 - 136781656 on Build GRCh38
Downstream RNA binding motif protein, X-linked RBMX HNRNPG, HNRPG, MRXS11, RBMXP1, RBMXRT, RNMX, hnRNP-G 21.4kb Sense Chr. - 136869194 - 136880780 on Build GRCh38

View Full Product Details