The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278556.1 | NP_001265485.1 | 1 - 2 | 155 | 132 |
NM_001287222.1 | NP_001274151.1 | 1 - 2 | 155 | 132 | |
GenBank mRNA | AK297309.1 | - | 1 - 2 | 86 | 132 |
AK311789.1 | - | 1 - 2 | 102 | 132 | |
BC067747.1 | - | 1 - 2 | 61 | 132 | |
BF434378.1 | - | 1 - 2 | 166 | 132 | |
BG620669.1 | - | 1 - 2 | 73 | 132 | |
BG723870.1 | - | 1 - 2 | 119 | 132 |