Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLCE
Entrez Gene ID
26035
Gene Name
glucuronic acid epimerase
Gene Aliases
HSEPI
Chromosome Location
Chr.15: 69160555 - 69272209 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005254298.3 XP_005254355.1 1 - 2 198 107

View Full Product Details