Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPL26
Entrez Gene ID
6154
Gene Name
ribosomal protein L26
Gene Aliases
DBA11, L26
Chromosome Location
Chr.17: 8377516 - 8383247 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000987.3 NP_000978.1 1 - 2 89 116
GenBank mRNA BI091167.1 - 1 - 2 89 116

View Full Product Details