Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBM27
Entrez Gene ID
54439
Gene Name
RNA binding motif protein 27
Gene Aliases
ARRS1, Psc1, ZC3H18, ZC3H20
Chromosome Location
Chr.5: 146203550 - 146289223 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005268466.1 XP_005268523.1 1 - 1 59 87
XM_005268467.1 XP_005268524.1 1 - 1 59 87
XM_005268468.1 XP_005268525.1 1 - 1 59 87
XM_006714787.1 XP_006714850.1 1 - 1 59 87
XM_011537661.1 XP_011535963.1 1 - 1 59 87
XM_011537662.1 XP_011535964.1 1 - 1 59 87
XM_017009587.1 XP_016865076.1 1 - 1 59 87
GenBank mRNA AK128738.1 - 1 - 1 59 87

View Full Product Details