Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSPO
Entrez Gene ID
23145
Gene Name
SCO-spondin
Gene Aliases
-
Chromosome Location
Chr.7: 149776042 - 149833965 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198455.2 NP_940857.2 57 - 58 8905 84

View Full Product Details