The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177663.1 | NP_001171134.1 | 33 - 34 | 7179 | 161 |
NM_001177664.1 | NP_001171135.1 | 33 - 34 | 7082 | 161 | |
NM_001177665.1 | NP_001171136.1 | 32 - 33 | 7352 | 161 | |
NM_018151.4 | NP_060621.3 | 34 - 35 | 7257 | 161 | |
XM_005246665.3 | XP_005246722.1 | 34 - 35 | 7137 | 161 | |
XM_011511395.2 | XP_011509697.1 | 33 - 34 | 4393 | 161 | |
XM_017004422.1 | XP_016859911.1 | 34 - 35 | 7206 | 161 | |
XM_017004423.1 | XP_016859912.1 | 33 - 34 | 7044 | 161 | |
XM_017004424.1 | XP_016859913.1 | 33 - 34 | 7074 | 161 | |
XM_017004425.1 | XP_016859914.1 | 32 - 33 | 7382 | 161 | |
GenBank mRNA | AK000323.1 | - | 5 - 6 | 1432 | 161 |
AL080129.1 | - | 5 - 6 | 1917 | 161 | |
AY584066.1 | - | 33 - 34 | 7096 | 161 | |
AY585745.1 | - | 34 - 35 | 7206 | 161 | |
AY727910.1 | - | 33 - 34 | 7179 | 161 | |
AY727911.1 | - | 33 - 34 | 7082 | 161 | |
AY727912.1 | - | 32 - 33 | 7352 | 161 | |
AY727913.1 | - | 34 - 35 | 7257 | 161 | |
U79263.1 | - | 4 - 5 | 699 | 161 |