The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145445.1 | NP_001138917.1 | 18 - 19 | 2723 | 64 |
XM_006724036.2 | XP_006724099.1 | 18 - 19 | 2692 | 64 | |
XM_017028416.1 | XP_016883905.1 | 18 - 19 | 2566 | 64 | |
XM_017028417.1 | XP_016883906.1 | 17 - 18 | 2527 | 64 | |
GenBank mRNA | AB032998.1 | - | 12 - 13 | 1711 | 64 |
AK025748.1 | - | 13 - 14 | 1742 | 64 | |
AK027017.1 | - | 4 - 5 | 534 | 64 | |
AL834304.1 | - | 18 - 19 | 2503 | 64 |