The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145122.1 | NP_001138594.1 | 18 - 19 | 1921 | 57 |
NM_001321270.1 | NP_001308199.1 | 18 - 19 | 1985 | 57 | |
XM_011532863.2 | XP_011531165.1 | 19 - 20 | 2082 | 57 | |
XM_011532864.2 | XP_011531166.1 | 19 - 20 | 2119 | 57 | |
XM_011532865.1 | XP_011531167.1 | 19 - 20 | 2023 | 57 | |
XM_011532866.2 | XP_011531168.1 | 17 - 18 | 1790 | 57 | |
GenBank mRNA | AK092257.1 | - | 18 - 19 | 1886 | 57 |
AK296910.1 | - | 20 - 21 | 1889 | 57 |