Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPN14
Entrez Gene ID
440854
Gene Name
calpain 14
Gene Aliases
-
Chromosome Location
Chr.2: 31173056 - 31233957 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145122.1 NP_001138594.1 21 - 22 2165 65
NM_001321270.1 NP_001308199.1 21 - 22 2229 65
XM_011532863.2 XP_011531165.1 22 - 23 2326 65
XM_011532864.2 XP_011531166.1 22 - 23 2363 65
XM_011532865.1 XP_011531167.1 22 - 23 2267 65
XM_011532866.2 XP_011531168.1 20 - 21 2034 65
GenBank mRNA AK092257.1 - 21 - 22 2130 65
AK296910.1 - 23 - 24 2133 65

View Full Product Details