The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198465.3 | NP_940867.2 | 12 - 13 | 1388 | 154 |
XM_006724632.3 | XP_006724695.1 | 12 - 13 | 1350 | 154 | |
XM_006724633.3 | XP_006724696.1 | 12 - 13 | 1350 | 154 | |
XM_006724634.3 | XP_006724697.1 | 12 - 13 | 1350 | 154 | |
XM_011530887.2 | XP_011529189.1 | 12 - 13 | 1142 | 154 | |
GenBank mRNA | BX538345.1 | - | 12 - 13 | 1390 | 154 |