Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIK3R6
Entrez Gene ID
146850
Gene Name
phosphoinositide-3-kinase regulatory subunit 6
Gene Aliases
C17orf38, HsT41028, p84 PIKAP, p87(PIKAP), p87PIKAP
Chromosome Location
Chr.17: 8802723 - 8870003 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010855.3 NP_001010855.1 11 - 12 1548 101
NM_001290211.1 NP_001277140.1 11 - 12 1916 101
NR_110865.1 10 - 11 1643 101
XM_011523677.2 XP_011521979.1 12 - 13 1561 101
XM_011523678.2 XP_011521980.1 12 - 13 1545 101
XM_011523680.2 XP_011521982.1 12 - 13 1561 101
XM_011523681.2 XP_011521983.1 11 - 12 1809 101
XM_011523683.2 XP_011521985.1 9 - 10 1246 101
XM_011523686.2 XP_011521988.1 3 - 4 695 101
XM_017024228.1 XP_016879717.1 12 - 13 1560 101
XM_017024229.1 XP_016879718.1 12 - 13 1560 101
XM_017024230.1 XP_016879719.1 12 - 13 1560 101
XM_017024231.1 XP_016879720.1 8 - 9 1737 101
GenBank mRNA AK091819.1 - 10 - 11 1645 101
AY753192.1 - 9 - 10 1308 101
BC110301.1 - 11 - 12 1899 101
BC125169.1 - 6 - 7 952 101

View Full Product Details