Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DEPDC1
Entrez Gene ID
55635
Gene Name
DEP domain containing 1
Gene Aliases
DEP.8, DEPDC1-V2, DEPDC1A, SDP35
Chromosome Location
Chr.1: 68474152 - 68497221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114120.2 NP_001107592.1 3 - 4 690 127
NM_017779.5 NP_060249.2 3 - 4 690 127
GenBank mRNA AB281187.1 - 3 - 4 546 127
AB281274.1 - 3 - 4 546 127
AJ278112.1 - 3 - 4 546 127
AK000361.1 - 3 - 4 551 127
AK225735.1 - 3 - 4 536 127
AK299324.1 - 3 - 4 551 127
BC003511.1 - 3 - 4 539 127
BC065304.1 - 3 - 4 493 127

View Full Product Details