The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152228.2 | NP_689414.1 | 1 - 2 | 195 | 76 |
XM_017002435.1 | XP_016857924.1 | 1 - 2 | 217 | 76 | |
XM_017002436.1 | XP_016857925.1 | 1 - 2 | 217 | 76 |