The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198461.3 | NP_940863.3 | 1 - 2 | 1318 | 159 |
XM_017003491.1 | XP_016858980.1 | 1 - 2 | 801 | 159 | |
GenBank mRNA | DQ020495.1 | - | 1 - 2 | 1318 | 159 |