The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276464.1 | NP_001263393.1 | 4 - 5 | 1051 | 71 |
NM_003822.4 | NP_003813.1 | 4 - 5 | 1220 | 71 | |
NM_205860.2 | NP_995582.1 | 5 - 6 | 1358 | 71 | |
XM_005245062.3 | XP_005245119.1 | 4 - 5 | 1485 | 71 | |
XM_011509380.1 | XP_011507682.1 | 6 - 7 | 1946 | 71 | |
XM_011509381.2 | XP_011507683.1 | 5 - 6 | 2810 | 71 | |
XM_011509382.1 | XP_011507684.1 | 5 - 6 | 1808 | 71 | |
XM_011509384.2 | XP_011507686.1 | 5 - 6 | 1191 | 71 | |
XM_017000904.1 | XP_016856393.1 | 4 - 5 | 1063 | 71 | |
GenBank mRNA | AB019246.1 | - | 5 - 6 | 1238 | 71 |
AF124247.1 | - | 5 - 6 | 1112 | 71 | |
AF146343.1 | - | 4 - 5 | 1181 | 71 | |
AK304344.1 | - | 4 - 5 | 1089 | 71 | |
AK304365.1 | - | 4 - 5 | 1051 | 71 | |
AK308446.1 | - | 4 - 5 | 1174 | 71 | |
AK316513.1 | - | 4 - 5 | 1287 | 71 | |
BC118571.1 | - | 4 - 5 | 1067 | 71 | |
BC118652.1 | - | 4 - 5 | 1067 | 71 | |
HQ692857.1 | - | 4 - 5 | 974 | 71 | |
U80251.1 | - | 4 - 5 | 1029 | 71 | |
U93553.1 | - | 5 - 6 | 1351 | 71 |