The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015103.2 | NP_055918.2 | 23 - 24 | 4297 | 112 |
XM_011512588.1 | XP_011510890.1 | 23 - 24 | 4212 | 112 | |
XM_011512589.1 | XP_011510891.1 | 20 - 21 | 3822 | 112 | |
XM_011512590.2 | XP_011510892.1 | 23 - 24 | 4212 | 112 | |
XM_011512592.1 | XP_011510894.1 | 11 - 12 | 1380 | 112 | |
GenBank mRNA | AB014520.2 | - | 23 - 24 | 4376 | 112 |
AK299425.1 | - | 2 - 3 | 129 | 112 | |
AY116661.1 | - | 23 - 24 | 4377 | 112 | |
BC150280.1 | - | 23 - 24 | 4376 | 112 | |
BU527983.1 | - | 5 - 6 | 554 | 112 |