The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015103.2 | NP_055918.2 | 26 - 27 | 4771 | 55 |
XM_011512588.1 | XP_011510890.1 | 26 - 27 | 4686 | 55 | |
XM_011512589.1 | XP_011510891.1 | 23 - 24 | 4296 | 55 | |
XM_011512592.1 | XP_011510894.1 | 14 - 15 | 1854 | 55 | |
GenBank mRNA | AB014520.2 | - | 26 - 27 | 4850 | 55 |
AK026388.1 | - | 2 - 3 | 268 | 55 | |
AK299425.1 | - | 5 - 6 | 603 | 55 | |
AY116661.1 | - | 26 - 27 | 4851 | 55 | |
BC003526.1 | - | 4 - 5 | 518 | 55 | |
BC011848.2 | - | 3 - 4 | 454 | 55 | |
BC150280.1 | - | 26 - 27 | 4850 | 55 |