The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052909.3 | NP_443141.3 | 6 - 7 | 1087 | 61 |
XM_006714446.3 | XP_006714509.1 | 8 - 9 | 2225 | 61 | |
XM_011513968.2 | XP_011512270.1 | 8 - 9 | 2225 | 61 | |
XM_011513969.2 | XP_011512271.1 | 8 - 9 | 2102 | 61 | |
XM_011513970.1 | XP_011512272.1 | 8 - 9 | 1814 | 61 | |
XM_011513971.2 | XP_011512273.1 | 8 - 9 | 2170 | 61 | |
XM_011513972.2 | XP_011512274.1 | 8 - 9 | 2226 | 61 | |
XM_011513973.1 | XP_011512275.1 | 8 - 9 | 2223 | 61 | |
XM_017009112.1 | XP_016864601.1 | 8 - 9 | 2226 | 61 | |
GenBank mRNA | AB067496.2 | - | 6 - 7 | 1087 | 61 |
BC008352.1 | - | 6 - 7 | 779 | 61 |