The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002081.2 | NP_002072.2 | 4 - 5 | 1101 | 109 |
XM_011510976.2 | XP_011509278.1 | 4 - 5 | 1002 | 109 | |
GenBank mRNA | BC045712.1 | - | 4 - 5 | 931 | 109 |
BC051279.1 | - | 4 - 5 | 1101 | 109 | |
BG825129.1 | - | 2 - 3 | 307 | 109 | |
X54232.1 | - | 4 - 5 | 1101 | 109 |