The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002081.2 | NP_002072.2 | 5 - 6 | 1236 | 105 |
XM_011510976.2 | XP_011509278.1 | 5 - 6 | 1137 | 105 | |
GenBank mRNA | AB209122.1 | - | 3 - 4 | 3196 | 105 |
AK096638.1 | - | 3 - 4 | 2795 | 105 | |
BC045712.1 | - | 5 - 6 | 1066 | 105 | |
BC051279.1 | - | 5 - 6 | 1236 | 105 | |
BG825129.1 | - | 3 - 4 | 442 | 105 | |
X54232.1 | - | 5 - 6 | 1236 | 105 |