The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002081.2 | NP_002072.2 | 7 - 8 | 1488 | 119 |
XM_011510976.2 | XP_011509278.1 | 7 - 8 | 1389 | 119 | |
GenBank mRNA | AB209122.1 | - | 5 - 6 | 3448 | 119 |
AK095397.1 | - | 6 - 7 | 2384 | 119 | |
BC051279.1 | - | 7 - 8 | 1488 | 119 | |
BG825129.1 | - | 5 - 6 | 117 | ||
X54232.1 | - | 7 - 8 | 1488 | 119 |