The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001224.4 | NP_001215.1 | 8 - 9 | 963 | 96 |
GenBank mRNA | CR541748.1 | - | 7 - 8 | 875 | 96 |
U13022.1 | - | 8 - 9 | 963 | 96 |