The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000547.5 | NP_000538.3 | 10 - 11 | 1856 | 83 |
NM_001206744.1 | NP_001193673.1 | 10 - 11 | 1851 | 83 | |
NM_175721.3 | NP_783652.1 | 9 - 10 | 1766 | 83 | |
NM_175722.3 | NP_783653.1 | 8 - 9 | 1247 | 83 | |
XM_011510379.2 | XP_011508681.1 | 9 - 10 | 1766 | 83 | |
XM_011510380.2 | XP_011508682.1 | 9 - 10 | 1766 | 83 | |
GenBank mRNA | AB208960.1 | - | 7 - 8 | 1552 | 83 |
AF439430.1 | - | 8 - 9 | 1246 | 83 | |
BC095448.1 | - | 10 - 11 | 1849 | 83 | |
CB986756.1 | - | - | 300 | 83 | |
J02969.1 | - | 10 - 11 | 1837 | 83 | |
M17755.2 | - | 10 - 11 | 1849 | 83 |