The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007254.3 | NP_009185.2 | 12 - 13 | 1234 | 64 |
GenBank mRNA | AF120499.1 | - | 12 - 13 | 1224 | 64 |
AF125807.1 | - | 11 - 12 | 1124 | 64 | |
AF126486.1 | - | 11 - 12 | 1124 | 64 | |
BC002519.2 | - | 11 - 12 | 1082 | 64 | |
BC033822.1 | - | 12 - 13 | 1230 | 64 |