The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012344.3 | NP_036476.1 | 3 - 4 | 1066 | 86 |
XM_005246156.3 | XP_005246213.1 | 2 - 3 | 795 | 86 | |
XM_006711876.3 | XP_006711939.1 | 3 - 4 | 1011 | 86 | |
XM_006711877.3 | XP_006711940.1 | 3 - 4 | 1002 | 86 | |
XM_017003738.1 | XP_016859227.1 | 3 - 4 | 1062 | 86 | |
GenBank mRNA | AY429107.1 | - | 3 - 4 | 992 | 86 |
BC022501.1 | - | 3 - 4 | 1027 | 86 | |
BC037776.2 | - | 4 - 5 | 980 | 86 | |
Y10148.1 | - | 3 - 4 | 1028 | 86 |