The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000574.4 | NP_000565.1 | 4 - 5 | 877 | 85 |
NM_001114752.2 | NP_001108224.1 | 4 - 5 | 877 | 85 | |
NM_001300902.1 | NP_001287831.1 | 4 - 5 | 877 | 85 | |
NM_001300903.1 | NP_001287832.1 | 4 - 5 | 877 | 85 | |
NM_001300904.1 | NP_001287833.1 | 4 - 5 | 877 | 85 | |
NR_125349.1 | 4 - 5 | 877 | 85 | ||
XM_017000467.1 | XP_016855956.1 | 4 - 5 | 711 | 85 | |
GenBank mRNA | AB240566.1 | - | 4 - 5 | 601 | 85 |
AB240567.1 | - | 4 - 5 | 601 | 85 | |
AB240568.1 | - | 4 - 5 | 601 | 85 | |
AB240569.1 | - | 4 - 5 | 616 | 85 | |
AB240570.1 | - | 4 - 5 | 616 | 85 | |
AF052110.1 | - | 4 - 5 | 818 | 85 | |
AK298549.1 | - | 5 - 6 | 579 | 85 | |
AK300621.1 | - | 3 - 4 | 477 | 85 | |
AK312448.1 | - | 4 - 5 | 672 | 85 | |
AY055758.1 | - | 2 - 3 | 198 | 85 | |
AY055759.1 | - | 2 - 3 | 198 | 85 | |
AY055760.1 | - | 2 - 3 | 198 | 85 | |
BC001288.1 | - | 4 - 5 | 662 | 85 | |
BT007159.1 | - | 4 - 5 | 583 | 85 | |
M15799.1 | - | 4 - 5 | 568 | 85 | |
M30142.1 | - | 4 - 5 | 648 | 85 | |
M31516.1 | - | 4 - 5 | 648 | 85 |