The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006516.2 | NP_006507.2 | 9 - 10 | 1809 | 76 |
GenBank mRNA | AB208987.1 | - | 9 - 10 | 1529 | 76 |
AF070544.1 | - | 6 - 7 | 837 | 76 | |
AK122999.1 | - | 7 - 8 | 1069 | 76 | |
AK292791.1 | - | 9 - 10 | 1495 | 76 | |
AK293306.1 | - | 9 - 10 | 1492 | 76 | |
AK296736.1 | - | 10 - 11 | 1327 | 76 | |
AK312403.1 | - | 9 - 10 | 1501 | 76 | |
BC118590.1 | - | 9 - 10 | 1430 | 76 | |
BC121804.1 | - | 9 - 10 | 1430 | 76 | |
BG682043.1 | - | 5 - 6 | 652 | 76 | |
K03195.1 | - | 9 - 10 | 1463 | 76 |