Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIE1
Entrez Gene ID
7075
Gene Name
tyrosine kinase with immunoglobulin like and EGF like domains 1
Gene Aliases
JTK14, TIE
Chromosome Location
Chr.1: 43300895 - 43323110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253357.1 NP_001240286.1 12 - 13 2026 86
NM_005424.4 NP_005415.1 12 - 13 2109 86
XM_005271163.1 XP_005271220.1 11 - 12 1923 86
XM_006710869.1 XP_006710932.1 12 - 13 2243 86
XM_017002207.1 XP_016857696.1 12 - 13 2052 86
GenBank mRNA AF217976.1 - 2 - 3 193 86
AK225560.1 - 12 - 13 1768 86
AK296735.1 - 7 - 8 1085 86
AK300399.1 - 12 - 13 1916 86
AK300623.1 - 12 - 13 1809 86
AK307639.1 - 5 - 6 777 86
BC038239.1 - 12 - 13 2011 86
EU826590.1 - 12 - 13 1932 86
X60957.1 - 12 - 13 1968 86

View Full Product Details