The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253357.1 | NP_001240286.1 | 18 - 19 | 3126 | 89 |
NM_005424.4 | NP_005415.1 | 18 - 19 | 3209 | 89 | |
XM_005271163.1 | XP_005271220.1 | 17 - 18 | 3023 | 89 | |
XM_006710869.1 | XP_006710932.1 | 18 - 19 | 3343 | 89 | |
GenBank mRNA | AF217976.1 | - | 8 - 9 | 1293 | 89 |
AK225560.1 | - | 18 - 19 | 2868 | 89 | |
AK296735.1 | - | 13 - 14 | 2185 | 89 | |
AK300399.1 | - | 18 - 19 | 3016 | 89 | |
AL833389.1 | - | 3 - 4 | 1661 | 89 | |
BC038239.1 | - | 18 - 19 | 3111 | 89 | |
X60957.1 | - | 18 - 19 | 3068 | 89 |