The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253357.1 | NP_001240286.1 | 7 - 8 | 1134 | 61 |
NM_005424.4 | NP_005415.1 | 7 - 8 | 1217 | 61 | |
XM_006710869.1 | XP_006710932.1 | 7 - 8 | 1351 | 61 | |
XM_017002207.1 | XP_016857696.1 | 7 - 8 | 1160 | 61 | |
GenBank mRNA | AK225560.1 | - | 7 - 8 | 1095 | 61 |
AK300399.1 | - | 7 - 8 | 1024 | 61 | |
BC038239.1 | - | 7 - 8 | 1119 | 61 | |
EU826590.1 | - | 7 - 8 | 1040 | 61 | |
X60957.1 | - | 7 - 8 | 1076 | 61 |