The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172251.2 | NP_758455.1 | 2 - 3 | 76 | |
GenBank mRNA | BC065273.1 | - | 2 - 3 | 309 | 76 |
BF346319.1 | - | 2 - 3 | 76 | ||
BM792364.1 | - | 2 - 3 | 76 | ||
BQ061807.1 | - | 2 - 3 | 76 |