Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEMA6B
Entrez Gene ID
10501
Gene Name
semaphorin 6B
Gene Aliases
SEM-SEMA-Y, SEMA-VIB, SEMAN, semaZ
Chromosome Location
Chr.19: 4542588 - 4581484 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032108.3 NP_115484.2 11 - 12 1378 77
XM_011527639.2 XP_011525941.1 11 - 12 1427 77
XM_011527640.2 XP_011525942.1 11 - 12 1170 77
XM_011527641.1 XP_011525943.1 11 - 12 1427 77
GenBank mRNA AB022433.1 - 10 - 11 1154 77
AF216389.1 - 10 - 11 1119 77
AF293363.1 - 10 - 11 1119 77
AK300037.1 - 11 - 12 1378 77
AY358939.1 - 10 - 11 1134 77
BC142617.1 - 11 - 12 1321 77
FR839673.1 - 10 - 11 1116 77

View Full Product Details